Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cIAP2 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA519385
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519385 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519385 Supplier Thermo Scientific Supplier No. PA519385

Goat Polyclonal Antibody

This antibody is predicted to react with human based on sequence homology.

cIAP2 Polyclonal antibody specifically detects cIAP2 in Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen cIAP2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene BIRC3
Gene Accession No. 0
Gene Alias AIP 2; AIP1; Api1; Api2; apoptosis inhibitor 1; apoptosis inhibitor 2; AW107670; baculoviral IAP repeat containing 3; baculoviral IAP repeat-containing 2; baculoviral IAP repeat-containing 3; baculoviral IAP repeat-containing protein 3; Birc2; Birc3; Cellular inhibitor of apoptosis 2; cIAP1; cIAP-1; cIAP2; C-IAP2; cIAP-2; H HIAP1; H MALT2; HAIP1; HIAP1; hIAP-1; HIAP2; IAP homolog; IAP homolog C; IAP1; IAP-1; IAP2; Inhibitor of apoptosis protein 1; MALT2; mammalian IAP homolog C; MIAP1; mIAP-1; MIAP2; MIHB; MIHC; RING finger protein 49; RING-type E3 ubiquitin transferase BIRC3; RNF49; TNFR2-TRAF signaling complex protein; TNFR2-TRAF-signaling complex protein 1
Gene Symbols BIRC3
Host Species Goat
Immunogen Synthetic peptide sequence (SVNNLEATSQP) corresponding to the internal amino acids of BIRC3
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 78971
Target Species Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.