Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CIAPIN1 Antibody (OTI5A6), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272072IR
Description
CIAPIN1 Monoclonal specifically detects CIAPIN1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CIAPIN1 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CIAPIN1 | |
Recombinant protein fragment corresponding to amino acids 97-312 of human CIAPIN1 (NP_064709) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI5A6 | |
Western Blot, Immunohistochemistry | |
Anamorsin, cytokine induced apoptosis inhibitor 1, Cytokine-induced apoptosis inhibitor 1, DRE2, Fe-S cluster assembly protein DRE2 homolog, predicted protein of HQ0915,2810413N20Rik, PRO0915 | |
Mouse | |
Affinity Purified | |
Apoptosis | |
57019.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction