Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CIAPIN1 Monoclonal Antibody (OTI1H4)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA527046
Description
CIAPIN1 Monoclonal Antibody for Western Blot, IHC (P)
CIAPIN1 is a cytokine-induced inhibitor of apoptosis with no relation to apoptosis regulatory molecules of the BCL2 or CASP families. Expression of CIAPIN1 is dependent on growth factor stimulation.
Specifications
CIAPIN1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6FI81 | |
CIAPIN1 | |
Human recombinant protein fragment corresponding to amino acids 97-312 of CIAPIN1 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1H4 | |
Unconjugated | |
CIAPIN1 | |
2810413N20Rik; AA617265; Anamorsin; anamorsin {ECO:0000255; AU021794; basic helix-loop-helix domain containing, class B, 2; basic helix-loop-helix domain containing, class B2; basic helix-loop-helix family member e40; basic helix-loop-helix family, member e40; BHLHB2; Bhlhe40; C130042M06Rik; CIAPIN1; Class B basic helix-loop-helix protein 2; class E basic helix-loop-helix protein 40; Clast5; CR8; CUA001; cytokine induced apoptosis inhibitor 1; cytokine response gene 8; cytokine-induced apoptosis inhibitor 1; cytokine-induced apoptosis inhibitor 1 {ECO:0000255; DEC1; differentially expressed in chondrocytes 1; differentially expressed in chondrocytes protein 1; differentiated embryo chondrocyte expressed gene 1; DRE2; E47 interaction protein 1; EIP1; eip1 (E47 interaction protein 1); enhancer-of-split and hairy-related protein 2; Fe-S cluster assembly protein DRE2 homolog; fe-S cluster assembly protein DRE2 homolog {ECO:0000255; HAMAP-Rule:MF_03115}; HLHB2; predicted protein of HQ0915; PRO0915; Sharp2; SHARP-2; Stimulated by retinoic acid gene 13 protein; Stra13; Stra14 | |
Mouse | |
Affinity chromatography | |
RUO | |
57019 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction