Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CIAPIN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA522321
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522321 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522321 Supplier Invitrogen™ Supplier No. PA522321
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: H1299. Predicted reactivity: Mouse (80%), Rat (82%), Chimpanzee (99%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CIAPIN1 is a cytokine-induced inhibitor of apoptosis with no relation to apoptosis regulatory molecules of the BCL2 or CASP families. Expression of CIAPIN1 is dependent on growth factor stimulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CIAPIN1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene CIAPIN1
Gene Accession No. Q6FI81
Gene Alias 2810413N20Rik; AA617265; Anamorsin; anamorsin {ECO:0000255; AU021794; basic helix-loop-helix domain containing, class B, 2; basic helix-loop-helix domain containing, class B2; basic helix-loop-helix family member e40; basic helix-loop-helix family, member e40; BHLHB2; Bhlhe40; C130042M06Rik; CIAPIN1; Class B basic helix-loop-helix protein 2; class E basic helix-loop-helix protein 40; Clast5; CR8; CUA001; cytokine induced apoptosis inhibitor 1; cytokine response gene 8; cytokine-induced apoptosis inhibitor 1; cytokine-induced apoptosis inhibitor 1 {ECO:0000255; DEC1; differentially expressed in chondrocytes 1; differentially expressed in chondrocytes protein 1; differentiated embryo chondrocyte expressed gene 1; DRE2; E47 interaction protein 1; EIP1; eip1 (E47 interaction protein 1); enhancer-of-split and hairy-related protein 2; Fe-S cluster assembly protein DRE2 homolog; fe-S cluster assembly protein DRE2 homolog {ECO:0000255; HAMAP-Rule:MF_03115}; HLHB2; predicted protein of HQ0915; PRO0915; Sharp2; SHARP-2; Stimulated by retinoic acid gene 13 protein; Stra13; Stra14
Gene Symbols CIAPIN1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CIAPIN1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57019
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.