Learn More
CIAPIN1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302809AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict
CIAPIN1 is a cytokine-induced inhibitor of apoptosis with no relation to apoptosis regulatory molecules of the BCL2 or CASP families. Expression of CIAPIN1 is dependent on growth factor stimulation.Specifications
CIAPIN1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q6FI81 | |
CIAPIN1 | |
Between 262 and 312 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
CIAPIN1 | |
CUA001, 2810413N20Rik, Anamorsin, DRE2, PRO0915, anamorsin, cytokine-induced apoptosis inhibitor 1, fe-S cluster assembly protein DRE2 homolog, predicted protein of HQ0915 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57019 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.