Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CIB1 Antibody (1D1), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP104279F
Description
CIB1 Monoclonal specifically detects CIB1 in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
CIB1 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
CIB1 | |
Recombinant human CIB1 (1-191aa) purified from E. coli | |
0.1 mL | |
Signal Transduction | |
10519 | |
Human | |
Purified |
ELISA | |
1D1 | |
ELISA | |
calcium and integrin binding 1 (calmyrin), Calcium- and integrin-binding protein, calmyrin, CIBcalcium and integrin binding protein, CIBP, DNA-dependent protein kinase interacting protein, DNA-PKcs-interacting protein, Kinase-interacting protein, KIP1, KIPcalcium and integrin binding, protein kinase interacting protein, PRKDCIP, SIP2-28calcium and integrin-binding protein 1, Snk interacting protein 2-28, SNK-interacting protein 2-28 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human CIB1 mAb, clone 1D1, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human CIB1 protein. | |
Store at 4C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction