Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CIB1 Mouse anti-Human, Clone: 5A1F5E12, Novus Biologicals™
SDP

Catalog No. p-7106199 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP147401SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP147401SS Supplier Novus Biologicals Supplier No. NBP147401SS

Mouse Monoclonal Antibody

CIB1 Monoclonal specifically detects CIB1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CIB1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 5A1F5E12
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000
Formulation Ascites with 0.03% Sodium Azide
Gene Alias calcium and integrin binding 1 (calmyrin), Calcium- and integrin-binding protein, calmyrin, CIBcalcium and integrin binding protein, CIBP, DNA-dependent protein kinase interacting protein, DNA-PKcs-interacting protein, Kinase-interacting protein, KIP1, KIPcalcium and integrin binding, protein kinase interacting protein, PRKDCIP, SIP2-28calcium and integrin-binding protein 1, Snk interacting protein 2-28, SNK-interacting protein 2-28
Gene Symbols CIB1
Host Species Mouse
Immunogen Purified recombinant fragment of CIB1 expressed in E. Coli.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10519
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.