Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CIB3 Monoclonal Antibody (OTI5F12), TrueMAB™, OriGene
SDP

Catalog No. 502543960
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3960 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3960 Supplier OriGene Supplier No. CF811077
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CIB3 Monoclonal antibody specifically detects CIB3 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen CIB3
Applications Western Blot
Classification Monoclonal
Clone OTI5F12
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene Stx3
Gene Accession No. Q08849, Q13277, Q64704
Gene Alias C730014M21Rik; calcium and integrin binding family member 3; calcium and integrin-binding family member 3; CIB3; DNA-dependent protein kinase catalytic subunit-interacting protein 3; Gm1107; Kinase-interacting protein 3; KIP 3; KIP3
Host Species Mouse
Immunogen Full length human recombinant protein of human STX3 (NP_004168) produced in HEK293T cell.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20908, 6809, 81802
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.