Learn More
CIC/Capicua Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301203A
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse
Transcriptional repressor which may play a role in development of the central nervous system (CNS).Specifications
| CIC/Capicua | |
| Polyclonal | |
| Unconjugated | |
| CIC | |
| Capicua , KIAA0306 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23152 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q96RK0 | |
| CIC | |
| Between 1150 and 1200 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.