Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CIC Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA146017

Catalog No. PA146017


Only null left
Add to Cart

Description

Description

PA1-46017 is not recommended for Western blot procedures.

This gene encodes a core component of the exosome. The mammalian exosome is required for rapid degradation of AU rich element-containing RNAs but not for poly(A) shortening. The association of this protein with the exosome is mediated by protein-protein interactions with ribosomal RNA-processing protein 42 and ribosomal RNA-processing protein 46.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CIC
Polyclonal
Unconjugated
CIC
1200010B10Rik; Capicua; capicua homolog; capicua homolog (Drosophila); capicua transcriptional repressor; CIC; KIAA0306; mKIAA0306; Protein capicua homolog; protein capicua homolog; LOW QUALITY PROTEIN: protein capicua homolog
Rabbit
Antigen Affinity Chromatography
RUO
23152, 308435, 484475, 71722
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunoprecipitation, Immunocytochemistry
1 mg/mL
PBS with 0.02% sodium azide
Q924A2, Q96RK0
CIC
A synthetic peptide made to a region within residues 1150-1200 of the human Capicua protein.
100 μg
Primary
Human, Mouse, Rat, Canine
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.