Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ciclopirox Olamine, USP, 97.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18601696
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle

C1486, 41621-49-2, C12H17NO2·C2H7NO

Spectrum™ Chemical Ciclopirox Olamine, USP is a synthetic antifungal agent for topical dermatologic treatment of superficial mycoses. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 41621-49-2
Molecular Formula C14H24N2O3
Molecular Weight (g/mol) 268.36
InChI Key MBRHNTMUYWQHMR-UHFFFAOYSA-N
IUPAC Name 2-aminoethan-1-ol; 6-cyclohexyl-1-hydroxy-4-methyl-1,2-dihydropyridin-2-one
SMILES NCCO.CC1=CC(=O)N(O)C(=C1)C1CCCCC1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Packaging Amber Glass Bottle
Quantity 500 g
Percent Purity 97.5-101.5%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.