Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CID 16020046
GREENER_CHOICESDP

Catalog No. 49595 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-595 5 mg
49-592-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 49-595 Supplier Tocris Bioscience™ Supplier No. 4959/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective GPR55 antagonist

CID 16020046 is a selective GPR55 antagonist. Inhibits LPI-induced Ca2+ signaling (IC50 = 0.21 μM in HEK-GPR55 cells), ERK1/2 phosphorylation and GPR55-mediated transcription factor activation. Displays weak inhibition of acetylcholinesterase, μ-opioid receptor, KCNH2 and hERG. Decreases LPI-induced GPR55 internalization. Reduces experimental intestinal inflammation in mice.

Specifications

Chemical Name or Material 4-[4,6-Dihydro-4-(3-hydroxyphenyl)-3-(4-methylphenyl)-6-oxopyrrolo[3,4-c]pyrazol-5(1H)-yl]benzoic acid
CAS 834903-43-4
Assay Percent Range >99%
Quantity 5 mg
Molecular Formula C25H19N3O4
Formula Weight Observed MW: 425.44
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.