Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CID Monoclonal Antibody (4F8), Active Motif™

Rat Monoclonal Antibody
$156.65 - $620.58
Specifications
Antigen | CID |
---|---|
Clone | 4F8 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3654
![]() ![]() |
Active Motif
61736 |
10 μg |
Each for $156.65
|
|
|||||
50-199-3653
![]() ![]() |
Active Motif
61735 |
100 μg |
Each for $620.58
|
|
|||||
Description
Centromeres are the differentiated chromosomal domains that specify the mitotic behavior of chromosomes. CENPA encodes a centromere protein which contains a histone H3 related histone fold domain that is required for targeting to the centromere. CENPA is proposed to be a component of a modified nucleosome or nucleosome-like structure in which it replaces 1 or both copies of conventional histone H3 in the (H3-H4)2 tetrameric core of the nucleosome particle.Specifications
CID | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
This antibody was raised against a recombinant protein corresponding to the N-terminus of drosophila CID. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles |
4F8 | |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rat | |
Drosophila | |
CenH3; CENP A; CENPA; CENP-A; Centromere autoantigen A; Centromere protein A; centromere protein A, 17kDa; centromere-specific histone; ggCENP-A; Histone H3-like centromeric protein A; RGD1563607 | |
IgG2a | |
Protein G | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title