Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CIDEA Polyclonal Antibody
CIDEA Polyclonal Antibody
Supplier: Thermo Scientific PA5115020
Description
CIDEA Polyclonal Antibody for Western Blot, ICC/IF, IHC (PFA)

Specifications
CIDEA | |
Polyclonal | |
Unconjugated | |
CIDEA | |
AW212747; cell death activator CIDE-A; Cell death-inducing DFFA-like effector A; cell death-inducing DNA fragmentation factor, alpha subunit-like effector A; cide a; Cidea; CIDE-A | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1149, 291541 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
O60543 | |
CIDEA | |
A synthesized peptide derived from human CIDEA(Accession O60543), corresponding to amino acid residues S20-T70. | |
100 μL | |
Primary | |
Human, Rat, Monkey | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction