Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CIDEB Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299269AF647
Description
CIDEB Polyclonal specifically detects CIDEB in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CIDEB | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
cell death-inducing DFFA-like effector bcell death activator CIDE-B | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of mouse CIDEB. | |
0.1 mL | |
Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication | |
27141 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction