Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CIDEB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520323
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520323 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520323 Supplier Invitrogen™ Supplier No. PA520323
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is mouse small intestine tissue lysate. PA5-20323 can be used with blocking peptide PEP-0443.

CIDEB activates apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CIDEB
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CIDEB
Gene Accession No. O70303, Q9UHD4
Gene Alias 1110030C18Rik; AI790179; Cell death activator CIDE-B; cell death-inducing DFFA-like effector B; cell death-inducing DNA fragmentation factor, alpha subunit-like effector B; CID; Cideb; CIDE-B; DFFA-like B
Gene Symbols CIDEB
Host Species Rabbit
Immunogen A peptide corresponding to 15 amino acids near the carboxy terminus of human CIDE-B.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12684, 27141, 364388
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.