Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CIDEC Mouse anti-Human, Clone:7C12F11, Abnova™

Catalog No. 89333093 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-093 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-093 Supplier Abnova Corporation Supplier No. MAB16728
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human CIDEC.

DNA fragmentation factor (DFF) induces the fragmentation of DNA associated with apoptosis. A novel family of cell death-inducing DFF45 (MIM 601882)-like effectors (CIDEs), including CIDEC, can also promote apoptosis (Liang et al., 2003 [PubMed 12429024]).[supplied by OMIM]

Specifications

Antigen CIDEC
Applications ELISA, Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 7C12F11
Conjugate Unconjugated
Description cell death-inducing DFFA-like effector c
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry
Immunocytochemistry (1:200-1:1000)
Flow Cytometry (1:200-1:400)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene CIDEC
Gene Alias CIDE-3, FLJ20871, Fsp27
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 53-141 of human CIDEC from E. coli.
Quantity 100 μg
Gene ID (Entrez) 63924
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.