Learn More
Invitrogen™ CIDEC Polyclonal Antibody

Description
Suggested positive control: antigen standard for CIDEC (transient overexpression lysate). Immunogen sequence: LLYPKSLSRH VSVRTS.
Specifications
Specifications
| Antigen | CIDEC |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | CIDEC |
| Gene Accession No. | P56198, Q5XI33, Q96AQ7 |
| Gene Alias | cell death activator CIDE-3; cell death inducing DFFA like effector c; cell death-inducing DFFA-like effector c; cell death-inducing DFFA-like effector protein C; CID; cide 3; CIDE3; CIDE-3; CIDE-3alpha; Cidec; fat specific gene 27; fat specific protein 27; fat-specific protein FSP27; Fat-specific protein FSP27 homolog; FPLD5; FSP27 |
| Gene Symbols | CIDEC |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.