Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CIDEC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110864
Change view
Click to view available options
Quantity:
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110864 100 μL
PIPA5110844 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5110864 Supplier Invitrogen™ Supplier No. PA5110864
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEYAMKSLSL LYPKSLSRHV SVRTSVVTQQ LLSEPSPKAP RARPCRVSTA DRSVRKGIMA YSLEDLLLK.

DNA fragmentation is one of the critical steps in apoptosis, which is induced by DNA fragmentation factor (DFF). DFF is composed of two subunits, a 40 kDa caspase-activated nuclease (DFF40) and a 45 kDa inhibitor (DFF45). Recently a novel family of cell-death-inducing DFF45-like effectors (CIDEs) has been identified. Human CIDE-3 is a novel member of CIDEs. There are 2 transcripts, CIDE-3 and CIDE-3alpha, were present in HepG2 and A375 cells. Consistent with its chromosome localization at 3p25, a region associated with high frequency loss of heterozygosity in many tumours, CIDE-3 may play an important role in prevention of tumorigenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CIDEC
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CIDEC
Gene Accession No. Q96AQ7
Gene Alias cell death activator CIDE-3; cell death inducing DFFA like effector c; cell death-inducing DFFA-like effector c; cell death-inducing DFFA-like effector protein C; CID; cide 3; CIDE3; CIDE-3; CIDE-3alpha; Cidec; fat specific gene 27; fat specific protein 27; fat-specific protein FSP27; Fat-specific protein FSP27 homolog; FPLD5; FSP27
Gene Symbols CIDEC
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human CIDEC. Recombinant protein control fragment (Product #RP-92098).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63924
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.