Learn More
CIDEC Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 122871AP150UL

Description
12287-1-AP recognizing CIDEC (calculated 28 kDa) as a 30 kDa and 45-50 kDa due to its posttranslational modification.
DNA fragmentation is one of the critical steps in apoptosis, which is induced by DNA fragmentation factor (DFF). DFF is composed of two subunits, a 40 kDa caspase-activated nuclease (DFF40) and a 45 kDa inhibitor (DFF45). Recently a novel family of cell-death-inducing DFF45-like effectors (CIDEs) has been identified. Human CIDE-3 is a novel member of CIDEs. There are 2 transcripts, CIDE-3 and CIDE-3alpha, were present in HepG2 and A375 cells. Consistent with its chromosome localization at 3p25, a region associated with high frequency loss of heterozygosity in many tumours, CIDE-3 may play an important role in prevention of tumorigenesis.Specifications
CIDEC | |
Polyclonal | |
Unconjugated | |
CIDEC | |
Cell death activator CIDE 3, CIDE 3, CIDEC, Fsp27 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
63924 | |
-20°C | |
Liquid |
Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96AQ7 | |
CIDEC | |
CIDEC Fusion Protein Ag2913 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.