Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cig2 Antibody (3A11/5), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308443R
Description
Cig2 Monoclonal antibody specifically detects Cig2 in S. pombe samples. It is validated for Western Blot, ELISASpecifications
Cig2 | |
Monoclonal | |
DyLight 550 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA | |
3A11/5 | |
50mM Sodium Borate | |
Bacterial S. pombe Cig2 Purified protein. | |
100 μg | |
Cell Cycle and Replication | |
S. pombe | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction