Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CIITA Antibody - (1-333 aa), Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177806
Description
CIITA Polyclonal specifically detects CIITA in Human samples. It is validated for Western Blot, ELISA.Specifications
CIITA | |
Polyclonal | |
Unconjugated | |
Antiserum with 0.01% Sodium Azide | |
C2TA, C2TAMHC class II transactivator type III, CIITAIV, class II, major histocompatibility complex, transactivator, MHC class II transactivator, MHC2TANLR family, acid domain containing, NLRA, nucleotide-binding oligomerization domain, leucine rich repeat and acid domaincontaining | |
Rabbit | |
123.51 kDa | |
0.1 mL | |
Primary | |
Human |
Western Blot, ELISA | |
90 mg/mL | |
Western Blot 1:500-1:3000, ELISA 1:5000-1:25000 | |
P33076 | |
CIITA | |
The immunogen used for this study was a bacterially produced recombinant FLAG-CIITA corresponding to amino acids 1 through 333 of the human protein. (Uniprot: P33076) | |
Delipidation and Defibrination | |
RUO | |
4261 | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction