Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CILP-1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB10041106
Description
CILP-1 Polyclonal specifically detects CILP-1 in Rat samples. It is validated for Western Blot, Peptide ELISA.Specifications
CILP-1 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
cartilage intermediate layer protein 1, cartilage intermediate layer protein 1 C1, cartilage intermediate layer protein 1 C2, cartilage intermediate layer protein, nucleotide pyrophosphohydrolase, Cartilage intermediate-layer protein, CILP-1, HsT18872 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Cross reacts with Rat. Expected from sequence similarity: Human, Mouse and Dog. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Western Blot 0.1 - 0.3 ug/ml, Immunohistochemistry-Paraffin 1:10 - 1:500, Peptide ELISA Detection limit 1:128000 | |
NP_003604.2 | |
CILP | |
Peptide with sequence C-NYRRTDHEDPRVK corresponding to internal region according to NP_003604.2. | |
0.1 mg | |
Immune System Diseases, Immunology | |
8483 | |
Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction