Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CILP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547748
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547748 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547748 Supplier Invitrogen™ Supplier No. PA547748
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs, less than 1% cross-reactivity with recombinant human CILP-1 C-terminal peptide is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Major alterations in the composition of the cartilage extracellular matrix occur in joint disease, such as osteoarthrosis. This gene encodes the cartilage intermediate layer protein activity. The full-length and the N-terminal domain of this protein was shown to function as an IGF-1 antagonist. An allelic variant of this gene has been associated with lumbar disc disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CILP
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene CILP
Gene Accession No. O75339
Gene Alias 9830114D16; C130036G17Rik; cartilage intermediate layer protein; cartilage intermediate layer protein 1; Cartilage intermediate layer protein 1 C1; cartilage intermediate layer protein 1 C2; cartilage intermediate layer protein, nucleotide pyrophosphohydrolase; cartilage intermediate-layer protein; Cilp; CILP-1; HGNC:1980; HsT18872; UNQ602/PRO1188
Gene Symbols CILP
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human CILP-1 N-Terminal Fragment Arg22-Arg720.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8483
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.