Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CIN85/SH3KBP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | CIN85/SH3KBP1 |
---|---|
Dilution | Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
CIN85/SH3KBP1 Polyclonal specifically detects CIN85/SH3KBP1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CIN85/SH3KBP1 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
30011 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:GDSPKIDLAGSSLSGILDKDLSDRSNDIDLEGFDSVVSSTEKLSHPTTSRPKATGRRPPSQSLTSSSLSSPDIFDSPSPEEDKEEHISLAHRGVDASKKTSKTVTISQVSDNKASLP | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Rabbit | |
Human, Mouse | |
c-Cbl-interacting protein, CD2-binding protein 3, CD2BP3, CIN85Cbl-interacting protein of 85 kDa, GIG10, HSB1, HSB-1, Human Src family kinase-binding protein 1, MIG18, migration-inducing gene 18, SH3 domain-containing kinase-binding protein 1, SH3-domain kinase binding protein 1, Src family kinase-binding protein 1, src-related kinase binding protein-1 | |
SH3KBP1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title