Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cinanserin hydrochloride, Tocris Bioscience™

Catalog No. p-4783352
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective 5-HT2 antagonist; also 3C-like proteinase inhibitor

5-HT2 antagonist. Also inhibits 3C-like proteinase (3CLpro; Mpro). Inhibits replication of SARS-CoV.

Chemical Identifiers

CAS 54-84-2
Molecular Formula C20H25ClN2OS
Molecular Weight (g/mol) 376.943
InChI Key LXGJPDKYMJJWRB-IERUDJENSA-N
Synonym cinanserin hydrochloride, cinnanserin, maptc, cinanserin hcl, unii-f65z32ki62, cinanserin hydrochloride usan, 2'-3-dimethylamino propyl thio cinnamanilide hydrochloride, 2'-3-dimethylamino propyl thio cinnamanilide monohydrochloride, 2-propenamide, n-2-3-dimethylamino propyl thio phenyl-3-phenyl-, monohydrochloride
PubChem CID 6433141
IUPAC Name (E)-N-[2-[3-(dimethylamino)propylsulfanyl]phenyl]-3-phenylprop-2-enamide;hydrochloride
SMILES CN(C)CCCSC1=CC=CC=C1NC(=O)C=CC2=CC=CC=C2.Cl

Specifications

Quantity 10 mg
Formula Weight 376.94
Percent Purity >99%
Chemical Name or Material Cinanserin hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.