Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CINAP Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298094MFV450
Description
CINAP Polyclonal antibody specifically detects CINAP in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
CINAP | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
CASK-interacting nucleosome assembly protein, CDA1CTCL-associated antigen se20-4, Cell division autoantigen 1, CINAPSE20-4, CTCL, CTCL tumor antigen se20-4, Cutaneous T-cell lymphoma-associated antigen se20-4, cutaneous T-cell lymphoma-associated tumor antigen se20-4, DENTTTSPY-like protein 2, differentially expressed nucleolar TGF-beta1 target, Differentially-expressed nucleolar TGF-beta1 target protein, HRIHFB2216, NP79, Nuclear protein of 79 kDa, SE204, testis-specific protein Y encoded-like 2, testis-specific Y-encoded-like protein 2, TSPX, TSPY-like 2 | |
Produced in rabbits immunized with E. coli-derived Human CINAP fragment. | |
0.1 mL | |
Apoptosis, Cell Biology, Neuroscience | |
64061 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction