Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-Cinnamaldehyde, 99%

Catalog No. AC110355000
Click to view available options
Quantity:
50 g
500 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 14371-10-9 | C9H8O | 132.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14371-10-9
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.16
MDL Number MFCD00007000
InChI Key KJPRLNWUNMBNBZ-QPJJXVBHSA-N
Synonym cinnamaldehyde, trans-cinnamaldehyde, cinnamic aldehyde, e-cinnamaldehyde, cinnamal, 3-phenylacrylaldehyde, zimtaldehyde, cinnamylaldehyde, 3-phenylpropenal, 2e-3-phenylprop-2-enal
PubChem CID 637511
ChEBI CHEBI:16731
SMILES O=C\C=C\C1=CC=CC=C1

Specifications

Melting Point -7.5°C
Color Yellow
Density 1.0500g/mL
Boiling Point 250.0°C to 252.0°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH=CHCHO
Refractive Index 1.6210 to 1.6230
Quantity 500 g
Beilstein 07,348
Merck Index 14,2297
Specific Gravity 1.05
Solubility Information Solubility in water: 1.1g/L (20°C) (kolbenmethode).
IUPAC Name (E)-3-phenylprop-2-enal
Formula Weight 132.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material trans-Cinnamaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.