Learn More
Cingulin Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303327AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Probably plays a role in the formation and regulation of the tight junction (TJ) paracellular permeability barrier.Specifications
Cingulin | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9P2M7 | |
CGN | |
Between 200 and 250 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
CGN | |
KIAA1319 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57530 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.