Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cinnamonitrile, 97%, predominantly trans
Click to view available options
Quantity:
25 g
100 g
Description
Cinnamonitrile is a parent compound whose derivatives can be applied as corrosion inhibitors for API J55 steel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cinnamonitrile is a parent compound whose derivatives can be applied as corrosion inhibitors for API J55 steel.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Air & light sensitive. Store in dark. Incompatible with oxidizing agents.
Cinnamonitrile is a parent compound whose derivatives can be applied as corrosion inhibitors for API J55 steel.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Air & light sensitive. Store in dark. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 18°C to 20°C |
| Density | 1.029 |
| Boiling Point | 253°C to 254°C |
| Flash Point | 126°C (259°F) |
| Refractive Index | 1.602 |
| Quantity | 25 g |
| Sensitivity | Air and light sensitive |
| Solubility Information | Soluble in alcohol. Insoluble in water. |
| Formula Weight | 129.16 |
| Percent Purity | 97% |
| Show More |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 217-552-5 |
| RTECSNumber | UD1440000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction