Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cinnamyl alcohol, 98% trans, ACROS Organics™

Click to view available options
Quantity:
5g
100g
250g
500g
Packaging:
Glass bottle
104-54-1
C9H10O
134.178
MFCD00002921
OOCCDEMITAIZTP-QPJJXVBHSA-N
cinnamyl alcohol,cinnamic alcohol,3-phenyl-2-propen-1-ol,3-phenylprop-2-en-1-ol,zimtalcohol,styryl carbinol,e-3-phenylprop-2-en-1-ol,3-phenylallyl alcohol,trans-cinnamyl alcohol,styryl alcohol
5315892
CHEBI:33227
(E)-3-phenylprop-2-en-1-ol
C1=CC=C(C=C1)C=CCO
104-54-1
C9H10O
134.178
MFCD00002921
OOCCDEMITAIZTP-QPJJXVBHSA-N
cinnamyl alcohol,cinnamic alcohol,3-phenyl-2-propen-1-ol,3-phenylprop-2-en-1-ol,zimtalcohol,styryl carbinol,e-3-phenylprop-2-en-1-ol,3-phenylallyl alcohol,trans-cinnamyl alcohol,styryl alcohol
5315892
CHEBI:33227
(E)-3-phenylprop-2-en-1-ol
C1=CC=C(C=C1)C=CCO
Packaging Glass bottle
Melting Point 31.0°C to 35.0°C
Boiling Point 258.0°C
Color White
Specific Gravity 1.04
Density 1.0400g/mL
Quantity 5g
Infrared Spectrum Authentic
Linear Formula C6H5CH=CHCH2OH
Beilstein 06, 570
Merck Index 15, 2303
Solubility Information Solubility in water: 1.8g/L (20°C). Other solubilities: soluble in glycerol, methanol, clearly soluble in 3 volumes 50% alcohol, freely soluble in alcohol, ether, and other, common organic solvents
Formula Weight 134.18
Physical Form Fused Low Melting Crystalline Solid
Percent Purity ≥97.5%
Chemical Name or Material Cinnamyl alcohol, 98%
Show More Show Less

GHS P Statement
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Contaminated work clothing should not be allowed out of the workplace.
Avoid breathing dust/fume/gas/mist/vapors/spray. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 203-212-3

RTECSNumber : GE2200000

TSCA : TSCA