Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cinnamyl alcohol, 98% trans

Catalog No. AC148522500
Click to view available options
Quantity:
5 g
100 g
250 g
500 g
Packaging:
Glass bottle

CAS: 104-54-1 | C9H10O | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-54-1
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00002921
InChI Key OOCCDEMITAIZTP-QPJJXVBHSA-N
Synonym cinnamyl alcohol, cinnamic alcohol, 3-phenyl-2-propen-1-ol, 3-phenylprop-2-en-1-ol, zimtalcohol, styryl carbinol, e-3-phenylprop-2-en-1-ol, 3-phenylallyl alcohol, trans-cinnamyl alcohol, styryl alcohol
PubChem CID 5315892
ChEBI CHEBI:33227
IUPAC Name (E)-3-phenylprop-2-en-1-ol
SMILES C1=CC=C(C=C1)C=CCO

Specifications

Melting Point 31.0°C to 35.0°C
Color White
Density 1.0400g/mL
Boiling Point 258.0°C
Flash Point 126°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula C6H5CH=CHCH2OH
Quantity 250 g
Beilstein 06, 570
Specific Gravity 1.04
Merck Index 15, 2303
Solubility Information Solubility in water: 1.8g/L (20°C). Other solubilities: soluble in glycerol, methanol, clearly soluble in 3 volumes 50% alcohol, freely soluble in alcohol, ether, and other, common organic solvents
Formula Weight 134.18
Percent Purity 98% trans
Physical Form Fused Low Melting Crystalline Solid
Chemical Name or Material Cinnamyl alcohol, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Contaminated work clothing should not be allowed out of the workplace.
Avoid breathing dust/fume/gas/mist/vapors/spray.
EINECSNumber 203-212-3
RTECSNumber GE2200000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.