Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cinnamylphosphonic Acid 98.0+%, TCI America™
SDP

Catalog No. C32041G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C32041G 1 g
C3204200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C32041G Supplier TCI America Supplier No. C32041G

Chemical Identifiers

CAS 146404-58-2
Molecular Formula C9H11O3P
Molecular Weight (g/mol) 198.158
MDL Number MFCD19105641
InChI Key AIWAKBITCLOITM-UHFFFAOYSA-N
Synonym 3-Phenyl-2-propenylphosphonic Acid
PubChem CID 69504832
IUPAC Name 3-phenylprop-2-enylphosphonic acid
SMILES C1=CC=C(C=C1)C=CCP(=O)(O)O

Specifications

Melting Point 173°C
Color White
Quantity 1 g
Formula Weight 198.16
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Cinnamylphosphonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.