Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CINP Antibody (1G10), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259486B
This item is not returnable.
View return policy
Description
CINP Monoclonal antibody specifically detects CINP in Human samples. It is validated for Western Blot, ELISASpecifications
CINP | |
Monoclonal | |
Biotin | |
CDK2-interacting protein, cyclin-dependent kinase 2 interacting protein, cyclin-dependent kinase 2-interacting protein, MGC849 | |
Recombinant human CINP (1-212aa) purified from E. coli | |
0.1 mL | |
Cell Cycle and Replication | |
51550 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA | |
1G10 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction