Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CINP Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299270AF647
Description
CINP Polyclonal specifically detects CINP in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| CINP | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| Alexa Fluor 647 | |
| CDK2-interacting protein, cyclin-dependent kinase 2 interacting protein, cyclin-dependent kinase 2-interacting protein, MGC849 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human CINP. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 51550 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction