Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CINP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112106
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112106 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112106 Supplier Invitrogen™ Supplier No. PA5112106
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CINP Polyclonal Antibody for Western Blot, ICC/IF

The function of this gene has not been reported. This gene contains a trinucleotide (ctt) repeat length polymorphism; alleles with either one or two repeat units have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CINP
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene CINP
Gene Accession No. Q9BW66
Gene Alias 1810047K05Rik; 2810452K22Rik; CDK2-interacting protein; Cinp; cyclin dependent kinase 2 interacting protein; cyclin-dependent kinase 2 interacting protein; cyclin-dependent kinase 2-interacting protein; MGC849; RGD1563395
Gene Symbols CINP
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the center region of the Human CINP.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51550
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.