Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS CIP 200™ Acid-Based Cleaner and Disinfectant

Catalog No. p-7221536 Shop All STERIS Corporation Products
Change view
Click to view available options
Volume (English):
1 gal.
5 gal.
30 gal.
55 gal.
250 gal.
300 gal.
Container Type:
Drum
Pail
Tote
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Container Type
17-463-032 1 gal. -
17-463-033 5 gal. Pail
17-463-034 30 gal. Drum
17-463-035 55 gal. Drum
17-463-036 - Tote
17-463-037 250 gal. Tote
17-463-038 300 gal. Tote
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 17-463-032 Supplier STERIS Supplier No. 1D2308
Only null left
Add to Cart
Edge
Add to Cart

Specially formulated to meet the unique cleaning demands found in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries that require an effective validatable detergent.

  • A proprietary blend of phosphoric acid, an advanced surfactant system and other performance-enhancing ingredients that provide multiple cleaning mechanisms
  • Low foaming detergent is ideal for use in CIP, COP, and manual applications for a variety of applications including derouging and passivation
  • Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports
  • Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics
  • Manufactured with biodegradable surfactants to support harmonization of cleaning processes
  • Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility

Specifications

Product Type Disinfectant
pH 1.8 to 2.2
Sterility Non-sterile
Appearance Clear
Composition Phosphoric Acid, Citric Acid
For Use With (Application) CIP, COP, Rouge Removal, and Passivation of Process Equipment
Quantity 4 Pk.
Solubility Complete
Volume (English) 1 gal.

Not yet available in the State of California

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.