Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS CIP 200™ Acid-Based Cleaner and Disinfectant

Catalog No. 17463038 Shop All STERIS Corporation Products
Click to view available options
Volume (English):
1 gal.
5 gal.
30 gal.
55 gal.
250 gal.
300 gal.
Container Type:
Drum
Pail
Tote

Specially formulated to meet the unique cleaning demands found in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries that require an effective validatable detergent.

  • A proprietary blend of phosphoric acid, an advanced surfactant system and other performance-enhancing ingredients that provide multiple cleaning mechanisms
  • Low foaming detergent is ideal for use in CIP, COP, and manual applications for a variety of applications including derouging and passivation
  • Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports
  • Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics
  • Manufactured with biodegradable surfactants to support harmonization of cleaning processes
  • Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility

Specifications

Product Type Disinfectant
pH 1.8 to 2.2
Sterility Non-sterile
Appearance Clear
Composition Phosphoric Acid, Citric Acid
Container Type Tote
For Use With (Application) CIP, COP, Rouge Removal, and Passivation of Process Equipment
Solubility Complete
Volume (English) 300 gal.

Not yet available in the State of California

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.