Learn More
CIP29 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303210A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat and Orangutan
This gene encodes a protein that is upregulated in response to various cytokines. The encoded protein may play a role in cell cycle progression. A translocation between this gene and the myeloid/lymphoid leukemia gene, resulting in expression of a chimeric protein, has been associated with acute myelomonocytic leukemia. Pseudogenes exist on chromosomes 7 and 8. Alternatively spliced transcript variants have been described.Specifications
CIP29 | |
Polyclonal | |
Unconjugated | |
SARNP | |
CIP29, HCC1, HSPC316, THO1, SAP domain-containing ribonucleoprotein, cytokine induced protein 29 kDa, cytokine-induced protein of 29 kDa, hepatocellular carcinoma 1, nuclear protein Hcc-1, proliferation associated cytokine-inducible protein CIP29, proliferation-associated cytokine-inducible protein CIP29 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84324 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P82979 | |
SARNP | |
Between 160 and 210 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.