Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CIP2A Antibody (2G10), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 17 publications
$206.50 - $499.50
Specifications
Antigen | CIP2A |
---|---|
Clone | 2G10 |
Concentration | 1.8 mg/mL |
Dilution | Western Blot 1:1000-1:2000, Flow Cytometry 1 ug per million cells, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/Immunofluorescence 1:40, Immunohistochemistry-Paraffin reported in scientific literature (PMID 25965834), KnockDown Validated |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11059722T
![]() |
Novus Biologicals
NB11059722SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB11059722
![]() |
Novus Biologicals
NB11059722 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
CIP2A Monoclonal specifically detects CIP2A in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
CIP2A | |
1.8 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cancer, Tumor Suppressors | |
Q8TCG1 | |
57650 | |
A synthetic peptide made to a C-terminal portion of the human CIP2A protein. [UniProt# Q8TCG1] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
2G10 | |
Western Blot 1:1000-1:2000, Flow Cytometry 1 ug per million cells, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/Immunofluorescence 1:40, Immunohistochemistry-Paraffin reported in scientific literature (PMID 25965834), KnockDown Validated | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Primate | |
Cancerous inhibitor of PP2A, CIP2A, FLJ12850, KIAA1524, MGC163436, p90, p90 autoantigen, protein CIP2A | |
CIP2A | |
IgG2b κ | |
Protein G purified | |
p90 autoantigen (2G10) |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title