Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CIP2A Antibody (2G10), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11059722MFV500
Description
CIP2A Monoclonal antibody specifically detects CIP2A in Human, Mouse, Rat, Primate samples. It is validated for Flow CytometrySpecifications
CIP2A | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Primate | |
Purified |
Flow Cytometry | |
2G10 | |
Flow Cytometry | |
Cancerous inhibitor of PP2A, CIP2A, FLJ12850, KIAA1524, MGC163436, p90, p90 autoantigen, protein CIP2A | |
A synthetic peptide made to a C-terminal portion of the human CIP2A protein. [UniProt# Q8TCG1] | |
0.1 mL | |
Cancer, Tumor Suppressors | |
57650 | |
Store at 4C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction