Learn More
CIP2A Mouse anti-Human, Mouse, Clone: 2A1-9A4, Bethyl Laboratories
Mouse Monoclonal Antibody
Supplier: Bethyl Laboratories, Inc A500010AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Inhibition of protein phosphatase 2A (PP2A) activity has been identified as a prerequisite for the transformation of human cells. The protein, designated Cancerous Inhibitor of PP2A (CIP2A), interacts directly with the oncogenic transcription factor c-Myc, inhibits PP2A activity toward c-Myc serine 62 (S62), and thereby prevents c-Myc proteolytic degradation. In addition to its function in c-Myc stabilization, CIP2A promotes anchorage-independent cell growth and in vivo tumor formation. The oncogenic activity of CIP2A is demonstrated by transformation of human cells by overexpression of CIP2A. Importantly, CIP2A is overexpressed in two common human maligcies, head and neck squamous cell carcinoma (HNSCC) and colon cancer.Specifications
CIP2A | |
Monoclonal | |
Unconjugated | |
CIP2A | |
Cancerous Inhibitor of Protein Phosphatase 2A , p90 | |
Mouse | |
Affinity chromatography | |
RUO | |
224171, 57650 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
2A1-9A4 | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q8BWY9, Q8TCG1 | |
CIP2A | |
Amino terminal one-third of the CIP2A recombit protein | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.