Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CIP2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579553
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579553 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579553 Supplier Invitrogen™ Supplier No. PA579553
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Mouse Testis Tissue, HELA whole cell.

Inhibition of protein phosphatase 2A (PP2A) activity has been identified as a prerequisite for the transformation of human cells. The protein, designated Cancerous Inhibitor of PP2A (CIP2A), interacts directly with the oncogenic transcription factor c-Myc, inhibits PP2A activity toward c-Myc serine 62 (S62), and thereby prevents c-Myc proteolytic degradation. In addition to its function in c-Myc stabilization, CIP2A promotes anchorage-independent cell growth and in vivo tumor formation. The oncogenic activity of CIP2A is demonstrated by transformation of human cells by overexpression of CIP2A. Importantly, CIP2A is overexpressed in two common human malignancies, head and neck squamous cell carcinoma (HNSCC) and colon cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CIP2A
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene Cip2a
Gene Accession No. Q8BWY9, Q8TCG1
Gene Alias AA408511; AU018569; C330027C09Rik; Cancerous inhibitor of PP2A; cancerous inhibitor of protein phosphatase 2A; Cip2a; Kiaa1524; p90; p90 autoantigen; p90 autoantigen homolog; Protein CIP2A; RGD1310335; RIKEN cDNA C330027C09 gene; similar to RIKEN cDNA C330027C09
Gene Symbols Cip2a
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human KIAA1524 (854-889aa EVQKAQLEGRLEEKESLVKLQQEELNKHSHMIAMIH).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 224171, 57650
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.