Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cip4 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10059833IR
Description
Cip4 Polyclonal antibody specifically detects Cip4 in Human samples. It is validated for Western BlotSpecifications
Cip4 | |
Polyclonal | |
Western Blot | |
cdc42-interacting protein 4, CIP4STOT, HSTP, Protein Felic, Salt tolerant protein, salt tolerator, STPTRIP-10, thyroid hormone receptor interactor 10, thyroid receptor interacting protein 10, Thyroid receptor-interacting protein 10, TR-interacting protein 10 | |
The immunogen recognized by this antibody maps to a region between residue 25 and 75 of human thyroid receptor interacting protein 10 (Cdc42-interacting protein 4) using the numbering given in entry AAM46851.1 (GeneID 9322). | |
0.1 mL | |
Cancer | |
9322 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction