Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CIPAR-1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297050MFV450
Description
CIPAR-1 Polyclonal antibody specifically detects CIPAR-1 in Human samples. It is validated for ELISASpecifications
CIPAR-1 | |
Polyclonal | |
ELISA | |
Castration-induced prostatic apoptosis-related protein 1, CIPAR-1, Prostatic androgen-repressed message 1 protein, Protein PARM-1 [Precursor] | |
Rabbits immunized with purified, recombinant Human CIPAR-1 (Accession#: AAH13294.1, Met1-Ser258) | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
25849 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction