Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cipar1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS7603R

Description
PARM1 may regulate TLP1 expression and telomerase activity, thus enabling certain prostatic cells to resist apoptosis.Specifications
Cipar1 | |
Polyclonal | |
Unconjugated | |
PARM1 | |
castration-induced prostatic apoptosis-related protein 1; Cipar1; DKFZP564O0823; PARM1; PARM-1; Prostate androgen-regulated mucin-like protein 1; prostatic androgen-regulated mucin-like protein 1; prostatic androgen-repressed message 1; prostatic androgen-repressed message-1; UNQ1879/PRO4322; WSC4; WSC4, cell wall integrity and stress response component 4 homolog | |
Rabbit | |
Protein A | |
RUO | |
25849 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q6UWI2 | |
PARM1 | |
KLH conjugated synthetic peptide derived from human Cipar1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction