Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CIRBP Monoclonal Antibody (6E12B3)
GREENER_CHOICE

Catalog No. PIMA531701
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531701 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531701 Supplier Invitrogen™ Supplier No. MA531701
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31701 has been tested in indirect ELISA.

CIRBP is cold-inducible mRNA binding protein that plays a protective role in the genotoxic stress response by stabilizing transcripts of genes involved in cell survival. It acts as a translational activator and seems to play an essential role in cold-induced suppression of cell proliferation. It binds specifically to the 3'-untranslated regions (3'-UTRs) of stress-responsive transcripts RPA2 and TXN. This protein acts as a translational repressor and promotes assembly of stress granules (SGs), when overexpressed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CIRBP
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 6E12B3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene CIRBP
Gene Accession No. Q14011
Gene Alias A18 hnRNP; A18HNRNP; CIRBP; CIRP; cold inducible RNA binding protein; cold inducible RNA-binding protein; cold-inducible RNA-binding protein; glycine-rich RNA binding protein; glycine-rich RNA-binding protein CIRP; R74941; testicular tissue protein Li 39
Gene Symbols CIRBP
Host Species Mouse
Immunogen Purified recombinant fragment of human CIRBP (AA: 1-90) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1153
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.