Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CIRBP Mouse anti-Human, Clone:6E12D5, Abnova™
Mouse monoclonal antibody raised against partial recombinant human CIRBP.
Supplier: Abnova Corporation MAB17922
Specifications
CIRBP | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Western Blot (1:100-1:500) The optimal working dilution should be determined by the end user. |
|
CIRBP | |
Mouse | |
100 μg | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
6E12D5 | |
cold inducible RNA binding protein | |
In PBS (0.05% sodium azide) | |
CIRP | |
Recombinant protein corresponding to amino acids 1-90 of human CIRBP. | |
1153 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction