Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec™ Circular Specimen Chucks for Leica, TBS, and Tanner Cryostats
SDP

Catalog No. p-200008106 Shop All Mopec Products
Click to view available options
Color:
Blue
Green
Orange
Pink
Purple
Red
White
Yellow
Length (Metric):
22 mm
25 mm
30 mm
40 mm
55 mm

Available in multiple sizes and colors for use with Leica, TBS, and Tanner Cryostats.

  • Round, aluminum specimen chucks
  • Suitable for Leica (except CM1950 model), TBS, and Tanner Cryostats

Specifications

For Use With (Equipment) Leica, TBS, and Tanner Cryostats
Length (Metric) 22 mm
Color Blue
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.