Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CIRH1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545139
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545139 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545139 Supplier Invitrogen™ Supplier No. PA545139
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ADVQSIAVAD QEDSFVVGTA EGTVFHFQLV PVTSNSSEKQ WVRTKPFQHH Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 79%; Rat: 100%.

CIRH1A encodes a WD40-repeat-containing protein that is localized to the nucleolus. Mutation of this gene causes North American Indian childhood cirrhosis, a severe intrahepatic cholestasis that results in transient neonatal jaundice, and progresses to periportal fibrosis and cirrhosis in childhood and adolescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CIRH1A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene UTP4
Gene Accession No. Q969X6
Gene Alias CIRH1A; Cirhin; cirrhosis, autosomal recessive 1A (cirhin); cirrhosis, autosomal recessive 1A (human); cPERP-E; Kiaa1988; Naic; Teg-292; testis expressed gene 292; testis-expressed gene 292; testis-expressed gene 292 protein; Tex292; U3 small nucleolar RNA-associated protein 4 homolog; UTP4; UTP4 small subunit (SSU) processome component; UTP4 small subunit processome component; UTP4, small subunit (SSU) processome component, homolog; UTP4, small subunit processome component
Gene Symbols UTP4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human CIRH1A.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84916
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.